Learn More
Potassium bisulfate, 98.5%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 207755000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium bisulfate | |
c.p. | |
White | |
500 g | |
HKO4S | |
MFCD00011404 | |
15, 7734 | |
Solubility in water: 490g/L (20°C). Other solubilities: decomposes in alcohol | |
[K+].OS([O-])(=O)=O | |
136.16 | |
136.16 | |
Analytical | |
2.32 |
214.0°C | |
7646-93-7 | |
Adhering Crystals | |
98.50% | |
KHSO4 | |
01,909 | |
potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c | |
CHKVPAROMQMJNQ-UHFFFAOYSA-M | |
potassium hydrogen sulfate | |
516920 | |
98.5% | |
Glass bottle |
Chemical Identifiers
7646-93-7 | |
136.16 | |
CHKVPAROMQMJNQ-UHFFFAOYSA-M | |
516920 | |
[K+].OS([O-])(=O)=O |
HKO4S | |
MFCD00011404 | |
potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c | |
potassium hydrogen sulfate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN
GHS Signal Word: Danger
EINECSNumber : 231-594-1
RTECSNumber : TS7200000
TSCA : TSCA
RUO – Research Use Only