missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium bromate, 99.5%, for analysis, Thermo Scientific Chemicals

$182.21 - $515.73

Chemical Identifiers

CAS 7758-01-2
Molecular Formula BrKO3
Molecular Weight (g/mol) 167.01
MDL Number MFCD00011359
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
Synonym potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
PubChem CID 23673461
ChEBI CHEBI:38211
IUPAC Name potassium;bromate
SMILES [O-]Br(=O)=O.[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC268392500
SDS View Documents
Thermo Scientific Chemicals
268392500
250 g Plastic bottle N/A
 
AC268390010
SDS View Documents
Thermo Scientific Chemicals
268390010
1 kg Plastic bottle N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
Specifications

Chemical Identifiers

7758-01-2
167.01
OCATYIAKPYKMPG-UHFFFAOYSA-M
23673461
potassium;bromate
BrKO3
MFCD00011359
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
CHEBI:38211
[O-]Br(=O)=O.[K+]

Specifications

350.0°C
White
Crystalline Powder
99.5% min. (Iodometry)
KBrO3
11,431
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
OCATYIAKPYKMPG-UHFFFAOYSA-M
potassium;bromate
23673461
167.01
Analytical
Plastic bottle
3.2700g/mL
7758-01-2
5 to 9 (5% soln. at 20°C)
250 g
BrKO3
MFCD00011359
15, 7738
Solubility in water: 70g/L (20°C). Other solubilities: practically insoluble in alcohol, soluble in boiling water
[O-]Br(=O)=O.[K+]
167.01
CHEBI:38211
99.5%
0.1% max. (105°C)
3.27
Potassium bromate, p.a.
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
May cause cancer.
May cause fire or explosion; strong oxidizer.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 231-829-8

RTECSNumber : EF8725000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only