missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium bromate, ACS, 99.8% min, Thermo Scientific Chemicals

$103.92 - $232.89

Chemical Identifiers

CAS 7758-01-2
Molecular Formula BrKO3
Molecular Weight (g/mol) 167.00
MDL Number MFCD00011359
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
Synonym potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
PubChem CID 23673461
ChEBI CHEBI:38211
IUPAC Name potassium;bromate
SMILES [O-]Br(=O)=O.[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4001322
View Documents
Thermo Scientific Chemicals
04001322
100 g
Each for $103.92
 
AA4001336
View Documents
Thermo Scientific Chemicals
04001336
500 g
Each for $232.89
 
Description

Description

Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7758-01-2
167.00
OCATYIAKPYKMPG-UHFFFAOYSA-M
23673461
potassium;bromate
BrKO3
MFCD00011359
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
CHEBI:38211
[O-]Br(=O)=O.[K+]

Specifications

370°C (decomposition)
Granules
99.8% min
MFCD00011359
Slightly soluble in alcohol but insoluble in acetone and ethanol.
[O-]Br(=O)=O.[K+]
167.00
CHEBI:38211
ACS Reagent
Potassium bromate
7758-01-2
100 g
BrKO3
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
OCATYIAKPYKMPG-UHFFFAOYSA-M
potassium;bromate
23673461
167
3.27 g/mL
Videos
Safety and Handling

Safety and Handling

P201-P202-P210-P220-P221-P260-P264b-P270-P271-P280-P283-P301+P310-P304+P340-P306+P360-P308+P313-P330-P370+P378q-P371+P380+P375-P501c

H271-H301-H335-H336-H351-H373

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 231-829-8

RTECSNumber : EF8725000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only