missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Carbonate, puriss p.a, ACS, ≥99.0% (T), Honeywell™ Fluka™
Puriss. p.a., ACS Reagent, anhydrous,≥99.0% (T)
$194.10 - $194.10
Chemical Identifiers
| CAS | 584-08-7 |
|---|---|
| Molecular Formula | CK2O3 |
| Molecular Weight (g/mol) | 138.21 |
| MDL Number | MFCD00011382 |
| InChI Key | BWHMMNNQKKPAPP-UHFFFAOYSA-L |
| Synonym | potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate |
| PubChem CID | 11430 |
| IUPAC Name | dipotassium;carbonate |
| SMILES | [K+].[K+].[O-]C([O-])=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 584-08-7 | |
| 138.21 | |
| BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
| 11430 | |
| [K+].[K+].[O-]C([O-])=O |
| CK2O3 | |
| MFCD00011382 | |
| potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate | |
| dipotassium;carbonate |
Specifications
| 891°C | |
| 584-08-7 | |
| CK2O3 | |
| MFCD00011382 | |
| 4267587 | |
| BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
| dipotassium;carbonate | |
| 11430 | |
| ≥99.0% (T) | |
| 2.430g/cm3 |
| Colorless or White | |
| 500 g | |
| K2CO3 | |
| NONH for all modes of transport | |
| potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate | |
| [K+].[K+].[O-]C([O-])=O | |
| 138.21 | |
| 138.21g/mol | |
| Poly bottle | |
| Potassium carbonate |
Safety and Handling
P261-P305 + P351 + P338
RTECSNumber : 209-529-3