missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Conductivity Standard, 300 μS/cm at 25°C, Ricca Chemical
Supplier: Ricca Chemical Company R58873501A
Specifications
| Potassium Chloride Conductivity Standard, 300μS/cm at 25°C | |
| 7447-40-7,7732-18-5 | |
| 99.98,0.02 | |
| Liquid | |
| MFCD00011360 | |
| Approximately 7 | |
| 100°C | |
| Miscible with water | |
| Conductivity Standard | |
| [Cl-].[K+] | |
| 74.55 | |
| 4873 | |
| Laboratory |
| Natural Poly Bottle | |
| 99.98,0.02 | |
| 1.0001 | |
| ClK | |
| Colorless | |
| 0°C | |
| Conductivity Meters and Probes | |
| Conductivity Standard | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 1 L | |
| CHEBI:32588 |
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| CHEBI:32588 | |
| [Cl-].[K+] |
| ClK | |
| MFCD00011360 | |
| 4873 | |
| potassium chloride |
Safety and Handling
ShelfLife : 24 months
Recommended Storage : Ambient