missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride (Crystalline/Certified ACS), Fisher Chemical™

Potassium chloride for calomel cells

$299.67 - $3352.25

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
P217-500
SDS View Documents Promotion Details
Thermo Fisher Scientific
P217500
500 g Poly Bottle
Each for $299.67
Case of 6 Each for $1,798.02
Only null left
 
P217-3
SDS View Documents Promotion Details
Fisher Chemical
P2173
3 kg Poly Bottle
Each for $1,188.50
Case of 4 Each for $4,754.00
Only null left
 
P217-10
SDS View Documents Promotion Details
Fisher Chemical
P21710
10 kg Poly Pail
Each for $3,352.25
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
potassium chloride
ClK
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
CHEBI:32588
[Cl-].[K+]

Specifications

1420°C
7447-40-7
6
500 g
ClK
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
[Cl-].[K+]
74.55
CHEBI:32588
99.0 to 100.5%
Pass Test
Potassium Chloride
770°C
White
Solid
99.0 to 100.5%
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
4873
74.55
Certified ACS
Poly Bottle
Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:0
Instability:1

SDS
missing translation for 'documents'

missing translation for 'documents'