missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Reference Solution D, 146.93 μS/cm at 25°C, 0.001 M, Ricca Chemical
$139.41 - $210.84
Chemical Identifiers
| CAS | 7447-40-7 |
|---|---|
| Molecular Formula | ClK |
| Molecular Weight (g/mol) | 74.55 |
| MDL Number | MFCD00011360 |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| PubChem CID | 4873 |
| ChEBI | CHEBI:32588 |
| IUPAC Name | potassium chloride |
| SMILES | [Cl-].[K+] |
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| CHEBI:32588 | |
| [Cl-].[K+] |
| ClK | |
| MFCD00011360 | |
| 4873 | |
| potassium chloride |
Specifications
| Natural Poly Bottle | |
| 0.001Molar | |
| 100.0,0.01 | |
| ClK | |
| Colorless | |
| 100°C | |
| Conductivity Standard | |
| [Cl-].[K+] | |
| 74.55 | |
| 4873 | |
| Potassium Chloride Reference Solution D, 146.93μS/cm (146.93μmho/cm) at 25°C |
| 7447-40-7,7732-18-5 | |
| 98.0,0.01 | |
| Liquid | |
| MFCD00011360 | |
| Approximately 7 | |
| Conductivity Standard | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 500 mL | |
| CHEBI:32588 |