Learn More
Potassium cyanate, 97%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 201111000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium cyanate | |
Pure | |
White | |
100 g | |
CKNO | |
MFCD00011411 | |
15, 7746 | |
Solubility in water: 750g/L (20°C). Other solubilities: insoluble in alcohol | |
[K+].[O-]C#N | |
81.12 | |
CHEBI:38904 | |
97% | |
Plastic bottle |
315.0°C | |
590-28-3 | |
Crystalline Powder and/or Chunks | |
97% | |
KOCN | |
01,1059 | |
potassium cyanate, kaliumcyanat, kocn, aero cyanate, cyanic acid, potassium salt, alicyanate, bulpur, potassiumcyanate, weedanol cyanol, caswell no. 688 | |
GKKCIDNWFBPDBW-UHFFFAOYSA-M | |
potassium;cyanate | |
11378442 | |
81.12 | |
Pure |
Chemical Identifiers
590-28-3 | |
81.12 | |
GKKCIDNWFBPDBW-UHFFFAOYSA-M | |
11378442 | |
potassium;cyanate |
CKNO | |
MFCD00011411 | |
potassium cyanate, kaliumcyanat, kocn, aero cyanate, cyanic acid, potassium salt, alicyanate, bulpur, potassiumcyanate, weedanol cyanol, caswell no. 688 | |
CHEBI:38904 | |
[K+].[O-]C#N |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 209-676-3
RUO – Research Use Only