missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Cyanide, 10% (w/v), Ricca Chemical
$238.53 - $286.03
Chemical Identifiers
CAS | 151-50-8 |
---|---|
Molecular Formula | CKN |
Molecular Weight (g/mol) | 65.116 |
InChI Key | NNFCIKHAZHQZJG-UHFFFAOYSA-N |
Synonym | potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french |
PubChem CID | 9032 |
IUPAC Name | potassium;cyanide |
SMILES | [C-]#N.[K+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
6040-16
|
Ricca Chemical Company
604016 |
500 mL | Amber Poly Bottle |
Each for $286.03
|
|
||||
R60400001B
|
Ricca Chemical Company
R60400001B |
1 L | Amber Poly Bottle |
Each for $238.53
|
|
||||
Chemical Identifiers
151-50-8 | |
65.116 | |
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french | |
potassium;cyanide |
CKN | |
NNFCIKHAZHQZJG-UHFFFAOYSA-N | |
9032 | |
[C-]#N.[K+] |
Specifications
Colorless | |
89.0,8.91 | |
CKN | |
NNFCIKHAZHQZJG-UHFFFAOYSA-N | |
potassium;cyanide | |
9032 | |
Aqueous Solution | |
Liquid | |
Potassium Cyanide |
151-50-8,7732-18-5 | |
93.0,9.28 | |
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french | |
[C-]#N.[K+] | |
65.116 | |
Amber Poly Bottle | |
500 mL | |
10% (w/v) |