missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Dichromate, 0.100 N (N/10), 0.0167 M (M/60), Ricca Chemical

Supplier:  Ricca Chemical Company 60605

 View more versions of this product

Catalog No. 6060-5


Explore available promotions

Description

Description

Specifications

Specifications

Potassium Dichromate
7778-50-9,7732-18-5
100.0,0.5
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
294.182
CHEBI:53444
ACS/EP/USP Volumetric Solution
Liquid
Orange
98.0,0.48
Cr2K2O7
KMUONIBRACKNSN-UHFFFAOYSA-N
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
24502
Cubitainer
20 L
0.100N,0.0167M

Chemical Identifiers

7778-50-9
294.182
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
CHEBI:53444
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Cr2K2O7
KMUONIBRACKNSN-UHFFFAOYSA-N
24502
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Videos
SDS
Documents

Documents

Promotions

Promotions