missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium dichromate, 99.5%, extra pure, Thermo Scientific Chemicals

Oxidation reagent

$106.63 - $167.38

Chemical Identifiers

CAS 7778-50-9
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.19
MDL Number MFCD00011367
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC197765000
SDS View Documents
Thermo Scientific Chemicals
197765000
500 g Plastic bottle
Each for $106.63
 
AC197760010
SDS View Documents
Thermo Scientific Chemicals
197760010
1 kg Plastic bottle
Each for $167.38
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

7778-50-9
294.19
KMUONIBRACKNSN-UHFFFAOYSA-N
24502
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Cr2K2O7
MFCD00011367
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
CHEBI:53444
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

500.0°C
7778-50-9
Crystals
99.4% min. (Iodometry)
K2Cr2O7
08,410; 09,383; 10,324; 12,405
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
KMUONIBRACKNSN-UHFFFAOYSA-N
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
24502
294.19
Extra Pure
Plastic bottle
398.0°C
Orange to Orange-Red
500 g
Cr2K2O7
MFCD00011367
15, 7748
Solubility in water: 125g/L (20°C). Other solubilities: 50 g/100 mL water (100°C), insoluble in alcohol
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
294.19
CHEBI:53444
99.5%
0.05% max. (110°C)
Potassium dichromate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficu

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major

GHS Signal Word: Danger

EINECSNumber : 231-906-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only