missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium dichromate, 99+%, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 424115000

 View more versions of this product

Catalog No. AC424115000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Specifications

500.0°C
398.0°C
Orange
500 g
Cr2K2O7
MFCD00011367
15, 7748
Solubility in water: 125g/L (20°C). Other solubilities: 50 g/100 mL water (100°C), insoluble in alcohol
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
294.19
CHEBI:53444
≥99%
0.05% max.
>500°C
Potassium dichromate
7778-50-9
Crystals
99% min. (Iodometry)
K2Cr2O7
08,410; 09,383; 10,324; 12,405
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
KMUONIBRACKNSN-UHFFFAOYSA-N
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
24502
294.19
ACS Reagent
Plastic bottle

Chemical Identifiers

7778-50-9
KMUONIBRACKNSN-UHFFFAOYSA-N
24502
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
MFCD00011367
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
CHEBI:53444
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficu

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major

GHS Signal Word: Danger

EINECSNumber : 231-906-6

RTECSNumber : HX7680000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only