missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Dihydrogen Phosphate/Di-sodium Hydrogen Phosphate, Certipur™, MilliporeSigma™
Supplier: Technidata 1019600001
Specifications
| Potassium Dihydrogen Phosphate/Di-sodium Hydrogen Phosphate | |
| Certified secondary standard reference material for pH measurement; directly traceable to primary SRM from NIST/PTB pH(S)= 6.865 /pH(S) = 7.413 (25°C) | |
| H2KO4P | |
| potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
| [K+].OP(O)([O-])=O | |
| 136.08 | |
| CHEBI:63036 | |
| Odorless | |
| 2 x 25 g |
| Colorless solid | |
| 7778-77-0 | |
| MFCD00011401 MFCD00147253 | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| potassium dihydrogen phosphate | |
| 516951 | |
| 138.08 | |
| Fibre case | |
| Solid |
Chemical Identifiers
| 7778-77-0 | |
| 136.08 | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| 516951 | |
| potassium dihydrogen phosphate |
| H2KO4P | |
| MFCD00011401 MFCD00147253 | |
| potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
| CHEBI:63036 | |
| [K+].OP(O)([O-])=O |