missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium diphosphate, 98%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 269230010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium diphosphate | |
Extra Pure | |
White | |
1 kg | |
K4O7P2 | |
MFCD00011393 | |
potassium pyrophosphate, potassium diphosphate, tkpp, tetrapotassium diphosphate, tetrapotassium pyrophosphate, diphosphoric acid, tetrapotassium salt, tetrapotassium diphosphorate, unii-b9w4019h5g, pyrophosphoric acid, tetrapotassium salt, tetrapotassium phosphonato phosphate | |
RYCLIXPGLDDLTM-UHFFFAOYSA-J | |
tetrapotassium;phosphonato phosphate | |
23740 | |
98% | |
Plastic bottle |
1090°C | |
7320-34-5 | |
Crystalline Powder | |
98% | |
K4O7P2 | |
15, 7783 | |
Solubility in water: soluble. Other solubilities: insoluble in alcohol | |
[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O | |
330.33 | |
330.34 | |
Extra Pure |
Chemical Identifiers
7320-34-5 | |
330.33 | |
RYCLIXPGLDDLTM-UHFFFAOYSA-J | |
23740 | |
[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O |
K4O7P2 | |
MFCD00011393 | |
potassium pyrophosphate, potassium diphosphate, tkpp, tetrapotassium diphosphate, tetrapotassium pyrophosphate, diphosphoric acid, tetrapotassium salt, tetrapotassium diphosphorate, unii-b9w4019h5g, pyrophosphoric acid, tetrapotassium salt, tetrapotassium phosphonato phosphate | |
tetrapotassium;phosphonato phosphate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS Signal Word: Warning
EINECSNumber : 230-785-7
RUO – Research Use Only