missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium diphosphate, 98%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 269230250

 View more versions of this product

Catalog No. AC269230250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium diphosphate
Extra Pure
White
25 g
K4O7P2
MFCD00011393
potassium pyrophosphate, potassium diphosphate, tkpp, tetrapotassium diphosphate, tetrapotassium pyrophosphate, diphosphoric acid, tetrapotassium salt, tetrapotassium diphosphorate, unii-b9w4019h5g, pyrophosphoric acid, tetrapotassium salt, tetrapotassium phosphonato phosphate
RYCLIXPGLDDLTM-UHFFFAOYSA-J
tetrapotassium;phosphonato phosphate
23740
98%
Glass Bottle
1090°C
7320-34-5
Crystalline Powder
98%
K4O7P2
15, 7783
Solubility in water: soluble. Other solubilities: insoluble in alcohol
[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O
330.33
330.34
Extra Pure

Chemical Identifiers

7320-34-5
330.33
RYCLIXPGLDDLTM-UHFFFAOYSA-J
23740
[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O
K4O7P2
MFCD00011393
potassium pyrophosphate, potassium diphosphate, tkpp, tetrapotassium diphosphate, tetrapotassium pyrophosphate, diphosphoric acid, tetrapotassium salt, tetrapotassium diphosphorate, unii-b9w4019h5g, pyrophosphoric acid, tetrapotassium salt, tetrapotassium phosphonato phosphate
tetrapotassium;phosphonato phosphate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS Signal Word: Warning

EINECSNumber : 230-785-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only