Learn More
Potassium ethyl xanthate, 97+%, Thermo Scientific Chemicals
$121.30 - $1782.77
Chemical Identifiers
CAS | 140-89-6 |
---|---|
Molecular Formula | C3H5KOS2 |
Molecular Weight (g/mol) | 160.29 |
MDL Number | MFCD00004931 |
InChI Key | JCBJVAJGLKENNC-UHFFFAOYSA-M |
Synonym | potassium ethylxanthate, potassium ethyl xanthate, ethylxanthic acid potassium salt, potassium xanthogenate, potassium xanthate, potassium o-ethyl carbonodithioate, potassium ethyl xanthogenate, potassium ethylxanthogenate, potassium o-ethyl dithiocarbonate, z 3 pesticide |
PubChem CID | 2735045 |
IUPAC Name | potassium;ethoxymethanedithioate |
SMILES | [K+].CCOC([S-])=S |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1145022
|
Thermo Scientific Chemicals
A1145022 |
100 g |
Each for $121.30
|
|
|||||
AAA1145036
|
Thermo Scientific Chemicals
A1145036 |
500 g |
Each for $433.79
|
|
|||||
AAA114500E
|
Thermo Scientific Chemicals
A114500E |
2500 g |
Each for $1,782.77
|
|
|||||
Description
Potassium ethyl xanthate is used as a reagent for extraction of metal ions into organic solvents and in the preparation of 2-mercapto benzimidazoles and 2-mercapto benzoxazoles by reacting with o-phenylenediamines and o-amino phenols respectively. It acts as a precursor in the production of unsymmetrical sulfides through cross-coupling reaction and as a floatation agent in the mining industry. It is involved in the determination of cadmium in water samples by substoichiometric radiochemical method and the adsorption of colloidal dye disperse Blue 3 onto pretreated polyester fabric.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPotassium ethyl xanthate is used as a reagent for extraction of metal ions into organic solvents and in the preparation of 2-mercapto benzimidazoles and 2-mercapto benzoxazoles by reacting with o-phenylenediamines and o-amino phenols respectively. It acts as a precursor in the production of unsymmetrical sulfides through cross-coupling reaction and as a floatation agent in the mining industry. It is involved in the determination of cadmium in water samples by substoichiometric radiochemical method and the adsorption of colloidal dye disperse Blue 3 onto pretreated polyester fabric.
Solubility
Soluble in water.
Notes
Light sensitive. Incompatible with strong acids, strong bases and strong oxidizing agents and water.
Chemical Identifiers
140-89-6 | |
160.29 | |
JCBJVAJGLKENNC-UHFFFAOYSA-M | |
2735045 | |
[K+].CCOC([S-])=S |
C3H5KOS2 | |
MFCD00004931 | |
potassium ethylxanthate, potassium ethyl xanthate, ethylxanthic acid potassium salt, potassium xanthogenate, potassium xanthate, potassium o-ethyl carbonodithioate, potassium ethyl xanthogenate, potassium ethylxanthogenate, potassium o-ethyl dithiocarbonate, z 3 pesticide | |
potassium;ethoxymethanedithioate |
Specifications
140-89-6 | |
1.558 g/mL | |
98% | |
MFCD00004931 | |
3596974 | |
potassium ethylxanthate, potassium ethyl xanthate, ethylxanthic acid potassium salt, potassium xanthogenate, potassium xanthate, potassium o-ethyl carbonodithioate, potassium ethyl xanthogenate, potassium ethylxanthogenate, potassium o-ethyl dithiocarbonate, z 3 pesticide | |
JCBJVAJGLKENNC-UHFFFAOYSA-M | |
potassium;ethoxymethanedithioate | |
2735045 | |
98% |
∽205°C (decomposition) | |
205°C (401°F) | |
C3H5KOS2 | |
100 g | |
14,7696 | |
Soluble in water. | |
[K+].CCOC([S-])=S | |
160.29 | |
160.3 | |
Potassium ethyl xanthate |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 205-439-3
RTECSNumber : FG1575000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only