missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Fluoride, Anhydrous (White Powder/Certified), Fisher Chemical™

Supplier:  Thermo Fisher Scientific O6113500

 View more versions of this product

Catalog No. O6113500



Explore available promotions

Description

Description

Specifications

Specifications

1505°C
860°C
White
Solid
≥98.5 %
KF
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
[F-].[K+]
58.10
CHEBI:66872
≥98.5%
Pass Test
Poly Bottle
Potassium Fluoride, Anhydrous
7789-23-3
8 to 9
500 g
FK
MFCD00011398
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium fluoride
522689
58.09
Certified
0.5% max.
2.48g/cm³

Chemical Identifiers

7789-23-3
58.10
NROKBHXJSPEDAR-UHFFFAOYSA-M
522689
potassium fluoride
FK
MFCD00011398
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
CHEBI:66872
[F-].[K+]
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 6.1, : Poison

SDS Documents

Documents

Promotions

Promotions