missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Guaiacolsulfonate Hemihydrate 98.0+%, TCI America™
Supplier: TCI America G016025G
Specifications
Potassium Guaiacolsulfonate Hemihydrate | |
White | |
MFCD00053298 | |
Guaiacolsulfonic Acid Potassium Salt, 4-Hydroxy-3-methoxybenzenesulfonic Acid Potassium Salt, Potassium 4-Hydroxy-3-methoxybenzenesulfonate | |
COC1=C(C=CC(=C1)S(=O)(=O)[O-])O.[K+] | |
242.286 | |
CHEBI:32033 | |
≥98.0% (HPLC,T) |
16241-25-1 | |
C7H7KO5S | |
25 g | |
QFRKWSPTCBGLSU-UHFFFAOYSA-M | |
potassium;4-hydroxy-3-methoxybenzenesulfonate | |
23679005 | |
242.29 | |
Crystalline Powder |
Chemical Identifiers
16241-25-1 | |
242.286 | |
QFRKWSPTCBGLSU-UHFFFAOYSA-M | |
23679005 | |
potassium;4-hydroxy-3-methoxybenzenesulfonate |
C7H7KO5S | |
MFCD00053298 | |
Guaiacolsulfonic Acid Potassium Salt, 4-Hydroxy-3-methoxybenzenesulfonic Acid Potassium Salt, Potassium 4-Hydroxy-3-methoxybenzenesulfonate | |
CHEBI:32033 | |
COC1=C(C=CC(=C1)S(=O)(=O)[O-])O.[K+] |
Safety and Handling
EINECSNumber : (9)-0331
TSCA : No