missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium hexacyanocobaltate(III), 90+%, Thermo Scientific Chemicals

$189.92 - $657.42

Chemical Identifiers

CAS 13963-58-1
MDL Number MFCD00011391
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA2312618
View Documents
Thermo Scientific Chemicals
02312618
50 g
Each for $189.92
 
AA2312630
View Documents
Thermo Scientific Chemicals
02312630
250 g
Each for $657.42
 
Description

Description

It is applied in formation of a xerogel via a cyanogel, the gel has potential use in solid-state gas sensors. It is also used as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13963-58-1
MFCD00011391

Specifications

(decomposition)
13963-58-1
50 g
MFCD00011391
Soluble in water and acetic acid solutions; Insoluble in alcohol
≥90%
Light sensitive
Potassium hexacyanocobaltate(III)
Brown to Yellow
Crystalline
K3[Co(CN)6]
14,7637
332.33
Odorless
1.906 g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H351-H302-H332-H317
Toxic in contact with skin.
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

P281-P308+P313-P501c

Danger

EINECSNumber : 237-742-1

RTECSNumber : GF9470000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only