missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%, Thermo Scientific Chemicals

Maximum level of impurities: Insoluble matter 0.01%, SO4 0.005%, Fe 0.001%, Cl 0.02%, Heavy Metals (as Pb) 0.001%

Supplier:  Thermo Scientific Chemicals 04198622

 View more versions of this product

Catalog No. AA4198622


Explore available promotions

Description

Description

Potassium hydrogen iodate acts as an oxidizing agent. Further, it serves as an intermediate for the synthesis of potassium iodate and also used as an analysis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium hydrogen iodate
13455-24-8
White
HI2KO6
ACAYDTMSDROWHW-UHFFFAOYSA-M
potassium;iodic acid;iodate
23700942
ACS
Powder
Primary Standard
100 g
99.95-100.05%
MFCD00011400
OI(=O)=O.[O-]I(=O)=O.[K+]
389.909
389.91
Soluble in water

Chemical Identifiers

13455-24-8
389.909
ACAYDTMSDROWHW-UHFFFAOYSA-M
potassium;iodic acid;iodate
HI2KO6
MFCD00011400
23700942
OI(=O)=O.[O-]I(=O)=O.[K+]
Videos
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H272-H314-H335

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 236-650-9

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only