missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 10% (w/v), Ricca Chemical

$84.13 - $406.69

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6130-4
Accessories View Documents
Ricca Chemical Company
61304
120 mL Natural Poly Bottle
Each for $84.13
Only null left
 
6130-16
Accessories View Documents
Ricca Chemical Company
613016
500 mL Natural Poly Bottle
Each for $165.66
Only null left
 
6130-32
SDS Accessories View Documents
Ricca Chemical Company
613032
1 L Natural Poly Bottle
Each for $238.22
Only null left
 
6130-1
SDS Accessories View Documents
Ricca Chemical Company
RI61301
4 L Natural Poly Bottle
Each for $406.69
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
CHEBI:32035
[OH-].[K+]
HKO
MFCD00003553
14797
potassium hydroxide

Specifications

Colorless
82.0,15.92
HKO
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
9.94 to 10.06% w/v
Natural Poly Bottle
Potassium Hydroxide Solution, 10% (w/v)
1310-58-3,7732-18-5
85.0,16.56
MFCD00003553
120 mL
[OH-].[K+]
56.11
CHEBI:32035
Laboratory
Liquid
Videos
Safety and Handling

Safety and Handling

ShelfLife : 36 months

SDS
missing translation for 'documents'

missing translation for 'documents'