missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide, 1N solution in water
CAS: 1310-58-3 | HKO | 56.11 g/mol
$62.11 - $104.18
Specifications
| Chemical Name or Material | Potassium hydroxide |
|---|---|
| Color | Colorless |
| Name Note | 1N solution in water |
| Boiling Point | 100.0°C |
| CAS | 1310-58-3,7732-18-5 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC278400010
|
Thermo Scientific Chemicals
278400010 |
1 L | Plastic bottle |
Each for $62.11
|
|
||||
|
AC278400025
|
Thermo Scientific Chemicals
278400025 |
2.5 L | Plastic bottle |
Each for $104.18
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Potassium hydroxide | |
| 1N solution in water | |
| 1310-58-3,7732-18-5 | |
| KOH | |
| MFCD00003553 | |
| 01,935; 02,346; 03,238; 04,409; 05,557; 06,486; 07,303; 08,415; 09,387; 11,439; 12,411; 13,258 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| 56.11 | |
| CHEBI:32035 | |
| 1.05 |
| Colorless | |
| 100.0°C | |
| HKO | |
| 1N, exact strength on the certificate of analysis | |
| 1814 | |
| 1.0500g/mL | |
| Solubility in water: soluble. | |
| [OH-].[K+] | |
| 14797 | |
| 56.11 |
RUO – Research Use Only