missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium hydroxide, ca. 85%, ACS reagent, Thermo Scientific Chemicals
$46.74 - $378.48
Chemical Identifiers
CAS | 1310-58-3 |
---|---|
Molecular Formula | HKO |
Molecular Weight (g/mol) | 56.11 |
MDL Number | MFCD00003553 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
PubChem CID | 14797 |
ChEBI | CHEBI:32035 |
IUPAC Name | potassium hydroxide |
SMILES | [OH-].[K+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC424140250
|
Thermo Scientific Chemicals
424140250 |
25 g |
Each for $46.74
|
|
|||||
AC424145000
|
Thermo Scientific Chemicals
424145000 |
500 g |
Each for $91.20
|
|
|||||
AC424140025
|
Thermo Scientific Chemicals
424140025 |
2.5 kg |
Each for $237.07
|
|
|||||
AC424140050
|
Thermo Scientific Chemicals
424140050 |
5 kg |
Each for $378.48
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
potassium hydroxide |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 | |
[OH-].[K+] |
Specifications
White | |
1310-58-3 | |
ca. 85% | |
KOH | |
01,935; 02,346; 03,238; 04,409; 05,557; 06,486; 07,303; 08,415; 09,387; 11,439; 12,411; 13,258 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
25 g | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
∼85% | |
Plastic Bottle | |
Potassium hydroxide, 85% |
1320.0°C | |
360.0°C | |
HKO | |
MFCD00003553 | |
15, 7761 | |
Solubility in water: 1120g/L (20°C). Other solubilities: 1780g/L water (100°C), soluble in ethanol (400g/L), soluble in ammonia, slightly soluble in ether | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
14797 | |
56.11 | |
ACS Reagent | |
Pellet |
Safety and Handling
Danger
EINECSNumber : 215-181-3
RTECSNumber : TT2100000
TSCA : TSCA
RUO – Research Use Only