Learn More
Potassium Hydroxide (Pellets/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific P2501
Specifications
13.5 | |
Yellow | |
1310-58-3 | |
≥85.0% | |
HKO | |
1813 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
14797 | |
56.11 | |
Certified ACS | |
Poly Bottle |
Potassium Hydroxide | |
1320°C | |
360°C | |
≥85.0% | |
MFCD00003553 | |
2.04g/cm³ | |
1 kg | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
≥85.0% | |
Pass Test | |
Solid |
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
potassium hydroxide |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 | |
[OH-].[K+] |
Safety and Handling
DANGER!
Emergency Overview
Causes severe burns by all exposure routes. Water reactive. Use personal protective equipment. Do not allow contact with water. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.
NFPA
Health:3
Flammability:0
Instability:1
DOTInformation : DOT Class 8, : Corrosive