missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Hydroxide Solution, 0.1N (N/10) (Certified), Fisher Chemical

Titrated at 25°C to pH 8.6 (phenolphthalein endpoint) against NIST™ potassium acid phthalate

Supplier:  Thermo Fisher Scientific HF10634

Catalog No. SP214-1



Explore available promotions

Description

Description

Specifications

Specifications

Potassium Hydroxide Solution
0.1N (N/10)
HKO
0.0995 to 0.1005N
1.3g/cm³
1 L
[OH-].[K+]
56.11
CHEBI:32035
Certified
Pass Test
Liquid
Colorless
1310-58-3,7732-18-5
MFCD00003553
1814
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
56.11
0.1N (N/10)
Poly Bottle

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]
Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Accessories
Promotions

Promotions