missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Hydroxide Solution, 0.1N (N/10) (In Methanol/Certified), Fisher Chemical™

Supplier:  Thermo Fisher Scientific FLSP2241

 View more versions of this product

Catalog No. SP224-1

Explore available promotions

Description

Description

Specifications

Specifications

13
Colorless
66.1°C
-100°C
MFCD00003553
2924
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
56.11
0.1N (N/10)
Glass Bottle
Liquid
Potassium Hydroxide Solution
0.1N (N/10), in methanol
1310-58-3,67-56-1,7732-18-5
HKO
0.0995 to 0.1005N
0.8g/cm³
1 L
[OH-].[K+]
56.11
CHEBI:32035
Certified
Pass Test
LOT 84L Potassium acid phthalate

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents

Documents

Accessories
Promotions

Promotions