missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing


Potassium Hydroxide Solution, 0.5N (N/2) (In Methanol/Certified), Fisher Chemical™
Titrated at 25°C to pH 8.2 (phenolphthalein endpoint) against standardized HCl
$310.08 - $310.08
Chemical Identifiers
CAS | 1310-58-3 |
---|---|
Molecular Formula | HKO |
Molecular Weight (g/mol) | 56.11 |
MDL Number | MFCD00003553 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
PubChem CID | 14797 |
ChEBI | CHEBI:32035 |
IUPAC Name | potassium hydroxide |
SMILES | [OH-].[K+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
SP222-1
|
Fisher Chemical
FLSP2221 |
1 L | Glass Bottle |
N/A
|
|
||||
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
potassium hydroxide |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 | |
[OH-].[K+] |
Specifications
Colorless | |
1310-58-3,67-56-1,7732-18-5 | |
HKO | |
0.497 to 0.500N | |
0.8g/cm³ | |
1 L | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
Certified | |
Pass Test | |
Liquid |
65.6°C | |
-100°C | |
MFCD00003553 | |
2924 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
14797 | |
56.11 | |
0.5N (N/2) | |
Glass Bottle | |
Potassium Hydroxide Solution, 0.5N (N/2), in methanol |
Safety and Handling


DOTInformation : DOT Class 3, : Flammable Liquid