missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Hydroxide Solution, 1N (Certified), Fisher Chemical
Titrated at 25°C to pH 8.6 (phenolphthalein endpoint) against NIST™ potassium acid phthalate
Supplier: Thermo Fisher Scientific FLSP208500
Specifications
13 | |
1N | |
HKO | |
0.995 to 1.005N | |
1.05g/cm³ | |
500 mL | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
Certified | |
Pass Test | |
LOT 84L Potassium acid phthalate |
Potassium Hydroxide Solution | |
1310-58-3,7732-18-5 | |
MFCD00003553 | |
1814 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
14797 | |
56.11 | |
1N | |
Poly Bottle | |
Liquid |
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
potassium hydroxide |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 | |
[OH-].[K+] |