missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Hydroxide Solution, 45% w/w (Certified), Fisher Chemical™
Supplier: Thermo Fisher Scientific SP2364
Specifications
12.0 | |
Colorless | |
1310-58-3,7732-18-5 | |
45 to 47 % | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
14797 | |
56.11 | |
Certified | |
Pass Test | |
2.6mmHg at 20°C |
Potassium Hydroxide Solution | |
45% w/w | |
-28.8°C | |
HKO | |
1.510g/cm³ | |
4 L | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
45 to 47% | |
45% | |
Poly Bottle | |
Liquid |
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
potassium hydroxide |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 | |
[OH-].[K+] |
Safety and Handling
DOTInformation : DOT Class 8, : Corrosive