missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Hydroxide Solution, 45% w/w (Certified), Fisher Chemical™

Supplier:  Fisher Chemical SP236500

 View more versions of this product

Catalog No. SP236-500




Description

Description

Specifications

Specifications

12.0
Colorless
1310-58-3,7732-18-5
45 to 47 %
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
56.11
Certified
Pass Test
2.6mmHg at 20°C
Potassium Hydroxide Solution
45% w/w
-28.8°C
HKO
1.510g/cm³
500 mL
[OH-].[K+]
56.11
CHEBI:32035
45 to 47%
45%
Poly Bottle
Liquid

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Accessories
Promotions

Promotions