Learn More
Thermo Scientific Chemicals Potassium indigotrisulfonate
$89.38 - $1207.96
Chemical Identifiers
CAS | 67627-18-3 |
---|---|
Molecular Formula | C16H7K3N2O11S3 |
Molecular Weight (g/mol) | 616.71 |
MDL Number | MFCD00013160 |
InChI Key | XOSMXDUITYWYGR-JRYLAINFSA-K |
Synonym | potassium indigotrisulfonate, indigotrisulfonic acid tripotassium salt, indigotrisulfonate potassium salt, indigotrisulfonic acid potassium salt, unii-5zza8n0abt, 5zza8n0abt, tripotassium indigotrisulfonate, tri-potassium indigotrisulfonate, tripotassium indigo-5,5',7-trisulfonate, tri-potassium indigo-5,5',7-trisulfonate |
PubChem CID | 6364606 |
IUPAC Name | tripotassium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5,7-disulfonate |
SMILES | [K+].[K+].[K+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1/NC2=C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)C1=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC215170010
|
Thermo Scientific Chemicals
215170010 |
1 g | Glass bottle |
Each for $89.38
|
|
||||
AC215170050
|
Thermo Scientific Chemicals
215170050 |
5 g | Glass bottle |
Each for $305.01
|
|
||||
AC215170250
|
Thermo Scientific Chemicals
215170250 |
25 g | Glass bottle |
Each for $1,207.96
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67627-18-3 | |
616.71 | |
XOSMXDUITYWYGR-JRYLAINFSA-K | |
6364606 | |
[K+].[K+].[K+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1/NC2=C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)C1=O |
C16H7K3N2O11S3 | |
MFCD00013160 | |
potassium indigotrisulfonate, indigotrisulfonic acid tripotassium salt, indigotrisulfonate potassium salt, indigotrisulfonic acid potassium salt, unii-5zza8n0abt, 5zza8n0abt, tripotassium indigotrisulfonate, tri-potassium indigotrisulfonate, tripotassium indigo-5,5',7-trisulfonate, tri-potassium indigo-5,5',7-trisulfonate | |
tripotassium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5,7-disulfonate |
Specifications
67627-18-3 | |
C16H7K3N2O11S3 | |
1 g | |
XOSMXDUITYWYGR-JRYLAINFSA-K | |
tripotassium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5,7-disulfonate | |
6364606 | |
Potassium indigotrisulfonate |
Glass bottle | |
MFCD00013160 | |
potassium indigotrisulfonate, indigotrisulfonic acid tripotassium salt, indigotrisulfonate potassium salt, indigotrisulfonic acid potassium salt, unii-5zza8n0abt, 5zza8n0abt, tripotassium indigotrisulfonate, tri-potassium indigotrisulfonate, tripotassium indigo-5,5',7-trisulfonate, tri-potassium indigo-5,5',7-trisulfonate | |
[K+].[K+].[K+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1/NC2=C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)C1=O | |
616.71 | |
616.71 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO – Research Use Only