missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate-Iodide, (1mL = 0.5mg SO32-), for Sulfite, Certified, 0.0125N ±0.0002N (0.002083M) , LabChem™

$70.90 - $622.00

Specifications

Chemical Name or Material Potassium Iodate-Iodide
Color Colorless to Yellow
Name Note (1mL = 0.5mg SO32-), for Sulfite
CAS Max % 99.49,0.44,0.04,0.03
CAS 7681-11-0,7758-05-6,144-55-8
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC196601
View Documents
LabChem
LC196601
500 mL
Each for $70.90
Only null left
 
LC196602
View Documents
LabChem
LC196602
1 L
Each for $113.63
Only null left
 
LC196604
missing translation for 'viewDocuments'
LabChem
LC196604
4 L
Each for $253.77
Only null left
 
LC196605
missing translation for 'viewDocuments'
LabChem
LC196605
20 L
Each for $622.00
Only null left
 
Description

Description

Specifications

Specifications

Potassium Iodate-Iodide
(1mL = 0.5mg SO32-), for Sulfite
7681-11-0,7758-05-6,144-55-8
0.0125N ±0.0002N (0.002083M)
Soluble in water
Certified
Traceable to NIST
Liquid
Colorless to Yellow
99.49,0.44,0.04,0.03
KIO3
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
214
Passes Test
Iodine vapor; Carbon dioxide; Carbon monoxide
Videos
SDS
Documents

Documents