missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodate solution, 0.017M, Honeywell™

Volumetric, 1/60 M KIO3 (0.1N)

Supplier:  Honeywell-Fluka 342731L

Catalog No. 6002496


Only null left
Explore available promotions

Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Specifications

Specifications

Potassium iodate solution
1 L
KIO3
NONH for all modes of transport
JLKDVMWYMMLWTI-UHFFFAOYSA-M
potassium;iodate
23665710
0.017M
5-6-7758
IKO3
MFCD00011406
potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
[O-]I(=O)=O.[K+]
214.00
214g/mol

Chemical Identifiers

5-6-7758
214.00
JLKDVMWYMMLWTI-UHFFFAOYSA-M
23665710
[O-]I(=O)=O.[K+]
IKO3
MFCD00011406
potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
potassium;iodate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions