missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Iodide, Certified, 20.0% (w/v) ±0.4% (w/v), LabChem™
Supplier: LabChem LC198001
Specifications
Potassium Iodide | |
Yellow | |
Liquid | |
IK | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
potassium;iodide | |
4875 | |
166 | |
Certified | |
Amber Poly | |
Iodine vapor |
7681-11-0,7732-18-5 | |
80,20 | |
500 mL | |
KI | |
Soluble in water | |
[K+].[I-] | |
166.003 | |
CHEBI:8346 | |
20.0% ± 0.4% (w/v) | |
Passes Test | |
1.15g/mL | |
1.15g/mL |
Chemical Identifiers
7681-11-0 | |
166.003 | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
CHEBI:8346 | |
[K+].[I-] |
IK | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
4875 | |
potassium;iodide |
Safety and Handling
GHS H Statement
Causes eye irritation.
GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
Recommended Storage : Room Temperature; Protect from light