missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Iodide, Certified, 20.0% (w/v) ±0.4% (w/v), LabChem™

Supplier:  LabChem LC198004

 View more versions of this product

Catalog No. LC198004


Explore available promotions

Description

Description

Specifications

Specifications

Potassium Iodide
Yellow
Liquid
IK
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium;iodide
4875
166
Certified
Poly Bottle
Iodine vapor
7681-11-0,7732-18-5
80,20
4 L
KI
Soluble in water
[K+].[I-]
166.003
CHEBI:8346
20.0% ±0.4% (w/v)
Passes Test
1.15g/mL
1.15g/mL

Chemical Identifiers

7681-11-0
166.003
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
CHEBI:8346
[K+].[I-]
IK
NLKNQRATVPKPDG-UHFFFAOYSA-M
4875
potassium;iodide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes eye irritation.

GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Recommended Storage : Room Temperature; Protect from light

SDS
Documents

Documents

Promotions

Promotions