missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Iodide, For Chlorine, Chlorine Dioxide, Certified, 0.50% (w/v) ±0.05%, LabChem™
$49.32 - $106.38
Specifications
Color | Clear, colorless |
---|---|
Format | Liquid |
Storage Requirements | Room Temperature; Protect from light |
Type | Potassium Iodide |
Specifications
Clear, colorless | |
Liquid | |
Room Temperature; Protect from light | |
Potassium Iodide |