missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium manganate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03950609
Description
It serves as an oxidizing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium manganate | |
10294-64-1 | |
10 g | |
MFCD00043089 | |
Soluble in water | |
[K+].[K+].[O-][Mn]([O-])(=O)=O | |
197.13 | |
197.13 |
190°C (decomposition) | |
Powder | |
K2MnO4 | |
potassium manganate, unii-5pi213d3us, dipotassium dioxido dioxo manganese, acmc-1buxv, manganic acid h2mno4 , dipotassium salt, potassium manganate 10g, 2k.mno4, manganic acid h2mno4 , potassium salt 1:2 | |
XKYFHUXZPRFUTH-UHFFFAOYSA-N | |
dipotassium;dioxido(dioxo)manganese | |
160931 |
Chemical Identifiers
10294-64-1 | |
197.13 | |
XKYFHUXZPRFUTH-UHFFFAOYSA-N | |
160931 | |
[K+].[K+].[O-][Mn]([O-])(=O)=O |
K2MnO4 | |
MFCD00043089 | |
potassium manganate, unii-5pi213d3us, dipotassium dioxido dioxo manganese, acmc-1buxv, manganic acid h2mno4 , dipotassium salt, potassium manganate 10g, 2k.mno4, manganic acid h2mno4 , potassium salt 1:2 | |
dipotassium;dioxido(dioxo)manganese |
Safety and Handling
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
DOTInformation : Hazard Class: 5.1; Packaging Group: III
EINECSNumber : 233-665-2
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only