Learn More
Potassium Nitrate (Cryst./Certified ACS), Fisher Chemical™
$403.42 - $2608.67
Chemical Identifiers
| CAS | 7757-79-1 |
|---|---|
| Molecular Formula | KNO3 |
| Molecular Weight (g/mol) | 101.10 |
| MDL Number | MFCD00011409 |
| InChI Key | FGIUAXJPYTZDNR-UHFFFAOYSA-N |
| Synonym | potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras |
| PubChem CID | 24434 |
| ChEBI | CHEBI:63043 |
| IUPAC Name | potassium nitrate |
| SMILES | [K+].[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
P263-100
|
Thermo Fisher Scientific
P263100 |
100 g | Poly Bottle |
N/A
|
N/A | ||||
|
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
|
P263-500
|
Fisher Chemical
P263500 |
500 g | Poly Bottle |
Each for $403.42
Case of 6 Each for $2,420.52
|
|
||||
|
P263-3
|
Fisher Chemical
P2633 |
3 kg | Poly Bottle |
Each for $2,608.67
Case of 4 Each for $10,434.68
|
|
||||
Chemical Identifiers
| 7757-79-1 | |
| 101.10 | |
| FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
| 24434 | |
| potassium nitrate |
| KNO3 | |
| MFCD00011409 | |
| potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras | |
| CHEBI:63043 | |
| [K+].[O-][N+]([O-])=O |
Specifications
| 400°C | |
| 7757-79-1 | |
| 6 to 8 | |
| 100 g | |
| KNO3 | |
| potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras | |
| [K+].[O-][N+]([O-])=O | |
| 101.10 | |
| CHEBI:63043 | |
| ≥99.0% | |
| Pass Test | |
| >400°C |
| 334°C | |
| White | |
| Solid | |
| ≥99.0 % | |
| MFCD00011409 | |
| FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
| potassium nitrate | |
| 24434 | |
| 101.1 | |
| Certified ACS | |
| Poly Bottle | |
| Potassium Nitrate |
Safety and Handling
DANGER!
Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. The toxicological properties have not been fully investigated. Wear personal protective equipment. Ensure adequate ventilation. Keep away from clothing and other combustible materials. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.
NFPA
Health:1
Flammability:1
Instability:2
DOTInformation : DOT Class 5.1, : Oxidizer