missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium nitrite, ACS, 96.0% min, Thermo Scientific Chemicals

$59.52 - $337.85

Chemical Identifiers

CAS 7758-09-0
Molecular Formula KNO2
Molecular Weight (g/mol) 85.10
MDL Number MFCD00011408
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
Synonym potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320
PubChem CID 516910
IUPAC Name potassium;nitrite
SMILES [K+].[O-]N=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1424509
View Documents
Thermo Scientific Chemicals
01424509
10 g
Each for $59.52
 
AA1424522
View Documents
Thermo Scientific Chemicals
01424522
100 g
Each for $144.27
 
AA1424536
View Documents
Thermo Scientific Chemicals
01424536
500 g
Each for $337.85
 
Description

Description

In analytical chemistry (testing for amino acids, cobalt, iodine, urea).Potassium nitrite is used as a food preservative, paint and coating additives and analytical reagent. It is used as a strong oxidizing agent as well as involved in the preparation of heat transfer salts. In addition, it is employed as corrosion inhibitors and anti-scaling agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7758-09-0
85.10
BXNHTSHTPBPRFX-UHFFFAOYSA-M
516910
[K+].[O-]N=O
KNO2
MFCD00011408
potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320
potassium;nitrite

Specifications

350°C (decomposition)
Solid
96.0% min
MFCD00011408
Soluble in water,hot alcohol,liquid NH3Soluble in water,alcohol and ammonia.
[K+].[O-]N=O
85.10
85.11
Hygroscopic
Potassium nitrite, For ACS analysis
7758-09-0
10 g
KNO2
potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320
BXNHTSHTPBPRFX-UHFFFAOYSA-M
potassium;nitrite
516910
ACS Reagent
1.915 g/mL
Videos
Safety and Handling

Safety and Handling

P201-P202-P210-P220-P221-P264b-P270-P281-P301+P310-P308+P313-P330-P370+P378q-P501c

H272-H301-H500

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 231-832-4

RTECSNumber : TT3750000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only