Learn More
Potassium osmium(VI) oxide dihydrate, 98+%, Thermo Scientific Chemicals
$130.01 - $391.95
Chemical Identifiers
CAS | 10022-66-9 |
---|---|
Molecular Formula | H4K2O6Os |
Molecular Weight (g/mol) | 368.453 |
MDL Number | MFCD00149919 |
InChI Key | DGODWNOPHMXOTR-UHFFFAOYSA-N |
Synonym | potassium osmate vi dihydrate, potassium dioxidodioxoosmium dihydrate, potassium osmate dihydrate, unii-5m2bwq2tyj, 5m2bwq2tyj, osmate oso42-, dipotassium, dihydrate, t-4, dipotassium tetrahydroxodioxoosmate, potassium osmate k2oso2 oh 4, potassium osmate k2oso4 , dihydrate |
PubChem CID | 53393272 |
IUPAC Name | dipotassium;dioxido(dioxo)osmium;dihydrate |
SMILES | O.O.[O-][Os](=O)(=O)[O-].[K+].[K+] |
Description
Potassium osmium(VI) oxide dihydrate acts as a catalyst for the oxidative cleavage of olefins to ketones and carboxylic acids. It is employed in an intramolecular amidohydroxylation of carbamoyloxytethered olefins providing hydroxy oxazolidinones. It is also used as a reagent in the synthesis of amphidinolide B, which shows potent antitumor activity against human solid and blood tumor cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10022-66-9 | |
368.453 | |
DGODWNOPHMXOTR-UHFFFAOYSA-N | |
53393272 | |
O.O.[O-][Os](=O)(=O)[O-].[K+].[K+] |
H4K2O6Os | |
MFCD00149919 | |
potassium osmate vi dihydrate, potassium dioxidodioxoosmium dihydrate, potassium osmate dihydrate, unii-5m2bwq2tyj, 5m2bwq2tyj, osmate oso42-, dipotassium, dihydrate, t-4, dipotassium tetrahydroxodioxoosmate, potassium osmate k2oso2 oh 4, potassium osmate k2oso4 , dihydrate | |
dipotassium;dioxido(dioxo)osmium;dihydrate |
Specifications
White | |
Powder | |
99+% | |
MFCD00149919 | |
Soluble in water. Slowly decomposes in aqueous solution forming the tetroxide. Insoluble in alcohol,etherSoluble in water. Insoluble in alcohol and ether. | |
O.O.[O-][Os](=O)(=O)[O-].[K+].[K+] | |
368.453 | |
368.43 (332.40 Anhydrous) | |
Purple powder |
10022-66-9 | |
0.25 g | |
H4K2O6Os | |
potassium osmate vi dihydrate, potassium dioxidodioxoosmium dihydrate, potassium osmate dihydrate, unii-5m2bwq2tyj, 5m2bwq2tyj, osmate oso42-, dipotassium, dihydrate, t-4, dipotassium tetrahydroxodioxoosmate, potassium osmate k2oso2 oh 4, potassium osmate k2oso4 , dihydrate | |
DGODWNOPHMXOTR-UHFFFAOYSA-N | |
dipotassium;dioxido(dioxo)osmium;dihydrate | |
53393272 | |
Hygroscopic | |
Potassium osmium(VI) oxide dihydrate |
Safety and Handling
P210-P220-P221-P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
H272-H290-H312+H332-H314-H335
DOTInformation : DOT Class: 5.1; Packaging Group: II
EINECSNumber : 243-247-1
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only