missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium oxalate hydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A122890I

 View more versions of this product

Catalog No. AAA122890I


Explore available promotions

Description

Description

Potassium oxalate hydrate is used as a chelating agent, a purifying agent in the pharmaceutical industry and a precipitating agent in rare-earth metal processing. It finds application as a grinding agent, as a rust remover in metal treatment and as a bleaching agent in the textile and wood industry. It is also used in waste water treatment and as a scale remover in automobile radiators. Furthermore, it is used as a reagent in analytical chemistry for the determination of calcium and other heavy metals as well as reducing agents in photography. It is also involved in the preparation of SBA-15 (nanostructured SiO2) supported vanadium oxide catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium oxalate hydrate is used as a chelating agent, a purifying agent in the pharmaceutical industry and a precipitating agent in rare-earth metal processing. It finds application as a grinding agent, as a rust remover in metal treatment and as a bleaching agent in the textile and wood industry. It is also used in waste water treatment and as a scale remover in automobile radiators. Furthermore, it is used as a reagent in analytical chemistry for the determination of calcium and other heavy metals as well as reducing agents in photography. It is also involved in the preparation of SBA-15 (nanostructured SiO2) supported vanadium oxide catalyst.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with halogens, ammonia, cyanides and heavy metals.
Specifications

Specifications

Potassium oxalate hydrate
>300°C
6 to 8.5
Odorless
MFCD00150033
3752576
14,7651
Soluble in water.
O.[K+].[K+].[O-]C(=O)C([O-])=O
184.23
166.22 (Anhydrous)
6487-48-5
2.13
99%
C2H2K2O5
5000 g
Hygroscopic
potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
QCPTVXCMROGZOL-UHFFFAOYSA-L
dipotassium;oxalate;hydrate
2724193
99%

Chemical Identifiers

6487-48-5
184.23
QCPTVXCMROGZOL-UHFFFAOYSA-L
2724193
O.[K+].[K+].[O-]C(=O)C([O-])=O
C2H2K2O5
MFCD00150033
potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
dipotassium;oxalate;hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

EINECSNumber : 209-506-8

RTECSNumber : RO2885000

TSCA : Yes

storageNote1 : Hygroscopic

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only