Learn More
Potassium oxalate monohydrate, ACS, 98.5-101.0%
CAS: 6487-48-5 | C2H2K2O5 | 184.23 g/mol
$57.53 - $443.10
Chemical Identifiers
| CAS | 6487-48-5 |
|---|---|
| Molecular Formula | C2H2K2O5 |
| Molecular Weight (g/mol) | 184.23 |
| MDL Number | MFCD00150033 |
| InChI Key | QCPTVXCMROGZOL-UHFFFAOYSA-L |
| Synonym | potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate |
| PubChem CID | 2724193 |
| IUPAC Name | dipotassium;oxalate;hydrate |
| SMILES | O.[K+].[K+].[O-]C(=O)C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA1345222
|
Thermo Scientific Chemicals
01345222 |
100 g |
Each for $57.53
|
|
|||||
|
AA1345236
|
Thermo Scientific Chemicals
01345236 |
500 g |
Each for $158.71
|
|
|||||
|
AA13452A3
|
Thermo Scientific Chemicals
013452A3 |
2 kg |
Each for $443.10
|
|
|||||
Description
Potassium oxalate monohydrate is used as a chelating agent; a purifying agent in pharmaceutical industry and a precipitating agent in rare-earth metal processing. It finds application as a grinding agent, a rust remover for metal treatment, a bleaching agent in textile and wood industry. It is also used in waste water treatment and as a scale remover in automobile radiators. Furthermore, it is used as a reagent in analytical chemistry for the determination of calcium and other heavy metals as well as a reducing agent in photography. It is also involved in the preparation of SBA-15 (nanostructured SiO2) supported vanadium oxide catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 6487-48-5 | |
| 184.23 | |
| QCPTVXCMROGZOL-UHFFFAOYSA-L | |
| 2724193 | |
| O.[K+].[K+].[O-]C(=O)C([O-])=O |
| C2H2K2O5 | |
| MFCD00150033 | |
| potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate | |
| dipotassium;oxalate;hydrate |
Specifications
| 6487-48-5 | |
| White | |
| 6 to 8.5 | |
| C2H2K2O5 | |
| MFCD00150033 | |
| 3752576 | |
| potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate | |
| QCPTVXCMROGZOL-UHFFFAOYSA-L | |
| dipotassium;oxalate;hydrate | |
| 2724193 | |
| 98.8 to 101.0% | |
| Powder or Granular Powder |
| >300°C | |
| 2.13 g/mL | |
| Odorless | |
| K2C2O4·H2O | |
| 100 g | |
| 14,7651 | |
| Soluble in water. | |
| O.[K+].[K+].[O-]C(=O)C([O-])=O | |
| 184.23 | |
| 184.23 (166.22 Anhydrous) | |
| ACS | |
| Potassium oxalate monohydrate |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
EINECSNumber : 209-506-8
RTECSNumber : RO2885000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only