missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium periodate, ACS, 99.8%-100.3%, Thermo Scientific Chemicals

$87.75 - $201.08

Chemical Identifiers

CAS 7790-21-8
Molecular Formula IKO4
Molecular Weight (g/mol) 229.999
MDL Number MFCD00011407
InChI Key FJVZDOGVDJCCCR-UHFFFAOYSA-M
Synonym potassium periodate, potassium metaperiodate, unii-q3loc6p66l, q3loc6p66l, periodic acid hio4 , potassium salt, potassium ion io4, ssmicroaea 1/4 o, acmc-20ajue, ssmicroaea 1/4 oni, periodic acid hio4 , potassium salt 1:1
PubChem CID 516896
IUPAC Name potassium;periodate
SMILES [O-]I(=O)(=O)=O.[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA2289614
View Documents Promotion Details
Thermo Scientific Chemicals
02289614
25 g
Each for $87.75
 
AA2289622
View Documents Promotion Details
Thermo Scientific Chemicals
02289622
100 g
Each for $201.08
 
Description

Description

For the colorimetric determination of Mn (oxidizes manganese compounds to permanganates), as oxidizing agentPotassium periodate is used as an oxidizing agent in organic synthesis and an analytical reagent for the determination of potassium and cerium. It is also used in colorimetric estimation of manganese. It reacts with ammonium cerium(IV) nitrate to prepare tris[trinitratocerium(IV)] paraperiodate which finds application as an oxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7790-21-8
229.999
FJVZDOGVDJCCCR-UHFFFAOYSA-M
516896
[O-]I(=O)(=O)=O.[K+]
IKO4
MFCD00011407
potassium periodate, potassium metaperiodate, unii-q3loc6p66l, q3loc6p66l, periodic acid hio4 , potassium salt, potassium ion io4, ssmicroaea 1/4 o, acmc-20ajue, ssmicroaea 1/4 oni, periodic acid hio4 , potassium salt 1:1
potassium;periodate

Specifications

582°C
Powder
99.8%-100.3%
MFCD00011407
Soluble in water.
[O-]I(=O)(=O)=O.[K+]
229.999
230
3.618
7790-21-8
25 g
IKO4
potassium periodate, potassium metaperiodate, unii-q3loc6p66l, q3loc6p66l, periodic acid hio4 , potassium salt, potassium ion io4, ssmicroaea 1/4 o, acmc-20ajue, ssmicroaea 1/4 oni, periodic acid hio4 , potassium salt 1:1
FJVZDOGVDJCCCR-UHFFFAOYSA-M
potassium;periodate
516896
ACS Reagent
Potassium periodate, For ACS analysis
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 232-196-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only