missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Permanganate (Crystalline/Certified ACS), Fisher Chemical™

Supplier:  Fisher Chemical P279500

 View more versions of this product

Catalog No. P279-500



Explore available promotions

Description

Description

Specifications

Specifications

Potassium Permanganate
7722-64-7
8
500 g
KMnO4
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
[O-][Mn](=O)(=O)=O.[K+]
158.032
158.03
Certified ACS
Glass Bottle
240°C
Brown
Powder/Solid
≥99 %
MFCD00011364
VZJVWSHVAAUDKD-UHFFFAOYSA-N
potassium;permanganate
516875
≥99%
Pass Test

Chemical Identifiers

7722-64-7
158.032
VZJVWSHVAAUDKD-UHFFFAOYSA-N
516875
[O-][Mn](=O)(=O)=O.[K+]
KMnO4
MFCD00011364
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
potassium;permanganate
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. May cause burns. May be harmful if swallowed. Use only under a chemical fume hood. Wear personal protective equipment. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Keep away from clothing and other combustible materials. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:2
Flammability:0
Instability:0

DOTInformation : DOT Class 5.1, : Oxidizer

SDS Documents

Documents

Promotions

Promotions