missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium phenyltrifluoroborate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1756806

 View more versions of this product

Catalog No. AAL1756806

Explore available promotions

Description

Description

Aryltrifluoroborate salts are more nucleophilic than the corresponding arylboronic acids and undergo ligand-free Pd-catalyzed cross-coupling reactions with arenediazonium tetrafluoroborates to give good yields of biaryls. Suzuki Cross-Coupling are conducted using organotrifluoroborates as a potent boronic acid surrogates. In the presence of Dicarbonyl­(2,4-pentanedionato)­rhodium(I)­, 39295, K aryl- and alkenyltrifluoroborates add to aldehydes and enones to give secondary alcohols and saturated ketones, respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aryltrifluoroborate salts are more nucleophilic than the corresponding arylboronic acids and undergo ligand-free Pd-catalyzed cross-coupling reactions with arenediazonium tetrafluoroborates to give good yields of biaryls. Suzuki Cross-Coupling are conducted using organotrifluoroborates as a potent boronic acid surrogates. In the presence of Dicarbonyl­(2,4-pentanedionato)­rhodium(I)­, 39295, K aryl- and alkenyltrifluoroborates add to aldehydes and enones to give secondary alcohols and saturated ketones, respectively.

Solubility
Soluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Specifications

Specifications

Potassium phenyltrifluoroborate
296°C to 301°C
MFCD01318172
7782070
Soluble in water.
[B-](C1=CC=CC=C1)(F)(F)F.[K+]
184.01
184
153766-81-5
C6H5BF3K
5 g
potassium phenyltrifluoroborate, potassium trifluoro phenyl borate, potassium trifluoro phenyl boranuide, pubchem11425, potassiumphenyltrifluoroborate, amtb100, phenyltrifluoropotassioboron v, potassium trifluoro phenyl borate 1-, potassium ion trifluoro phenyl boranuide, potassium tris fluoranyl-phenyl-boranuide
DVAFPKUGAUFBTJ-UHFFFAOYSA-N
potassium;trifluoro(phenyl)boranuide
23675248
98%

Chemical Identifiers

153766-81-5
184.01
DVAFPKUGAUFBTJ-UHFFFAOYSA-N
23675248
[B-](C1=CC=CC=C1)(F)(F)F.[K+]
C6H5BF3K
MFCD01318172
potassium phenyltrifluoroborate, potassium trifluoro phenyl borate, potassium trifluoro phenyl boranuide, pubchem11425, potassiumphenyltrifluoroborate, amtb100, phenyltrifluoropotassioboron v, potassium trifluoro phenyl borate 1-, potassium ion trifluoro phenyl boranuide, potassium tris fluoranyl-phenyl-boranuide
potassium;trifluoro(phenyl)boranuide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only