missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate, dibasic, 99+%, for analysis, anhydrous
Potassium phosphate, dibasic, 99+%, for analysis, anhydrous, Quantity: 5kg, Packaging: Plastic drum, CAS: 7758-11-4, White, Grade: Analysis, Melting Point: 340.0 deg.C, Molecular Formula: HK2O4P, Molecular Weight: 174.18, Percent Purity: 99+%, pH: 8.5 to 9.6 (5% soln. at 20 deg.C) | CAS: 7758-11-4 | HK2O4P | 174.18 g/mol
$150.00 - $1451.58
Chemical Identifiers
| CAS | 11-4-7758 |
|---|---|
| Molecular Formula | HK2O4P |
| Molecular Weight (g/mol) | 174.18 |
| InChI Key | ZPWVASYFFYYZEW-UHFFFAOYSA-L |
| Synonym | dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate |
| PubChem CID | 24450 |
| ChEBI | CHEBI:32031 |
| IUPAC Name | dipotassium;hydrogen phosphate |
| SMILES | OP(=O)([O-])[O-].[K+].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC215472500
|
Thermo Scientific Chemicals
215472500 |
250 g | Plastic bottle |
Each for $150.00
|
|
||||
|
AC215470010
|
Thermo Scientific Chemicals
215470010 |
1 kg | Plastic bottle |
Each for $409.25
|
|
||||
|
AC215470050
|
Thermo Scientific Chemicals
215470050 |
5 kg | Plastic drum |
Each for $1,451.58
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisChemical Identifiers
| 11-4-7758 | |
| 174.18 | |
| dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
| CHEBI:32031 | |
| OP(=O)([O-])[O-].[K+].[K+] |
| HK2O4P | |
| ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
| 24450 | |
| dipotassium;hydrogen phosphate |
Specifications
| 340.0°C | |
| White | |
| Fine Crystalline Powder | |
| 99+% | |
| K2HPO4 | |
| dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
| ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
| dipotassium;hydrogen phosphate | |
| 24450 | |
| 174.18 | |
| Analytical | |
| Plastic bottle |
| 11-4-7758 | |
| 8.5 to 9.6 (5% soln. at 20°C) | |
| 250 g | |
| HK2O4P | |
| 15,7779 | |
| Solubility in water: 1600g/L (20°C). Other solubilities: slightly soluble in alcohol | |
| OP(=O)([O-])[O-].[K+].[K+] | |
| 174.18 | |
| CHEBI:32031 | |
| ≥99% | |
| 1% (130°C) max. | |
| Potassium phosphate, dibasic |
Safety and Handling
EINECSNumber : 231-834-5
RUO – Research Use Only