missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Phosphate Dibasic Trihydrate, (White, crystalline powder/Reagent Grade), MP Biomedicals

$375.14 - $1269.01

Chemical Identifiers

CAS 16788-57-1
Molecular Formula H7K2O7P
Molecular Weight (g/mol) 228.22
MDL Number MFCD00149926
InChI Key XQGPKZUNMMFTAL-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate trihydrate, dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
PubChem CID 16217523
IUPAC Name dipotassium trihydrate hydrogen phosphate
SMILES O.O.O.[K+].[K+].OP([O-])([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19143201
View Documents
MP Biomedicals Inc
0219143201
1 kg
Each for $375.14
 
ICN19143205
View Documents
MP Biomedicals Inc
0219143205
5 kg
Each for $1,269.01
 
Description

Description

Specifications

Chemical Identifiers

16788-57-1
228.22
XQGPKZUNMMFTAL-UHFFFAOYSA-L
16217523
O.O.O.[K+].[K+].OP([O-])([O-])=O
H7K2O7P
MFCD00149926
dipotassium hydrogen phosphate trihydrate, dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
dipotassium trihydrate hydrogen phosphate

Specifications

White
Crystalline Powder
H7K2O7P
dipotassium hydrogen phosphate trihydrate, dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
O.O.O.[K+].[K+].OP([O-])([O-])=O
228.22
228.2
Potassium Phosphate Dibasic Trihydrate
16788-57-1
1 kg
MFCD00149926
XQGPKZUNMMFTAL-UHFFFAOYSA-L
dipotassium trihydrate hydrogen phosphate
16217523
Reagent
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'