missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Potassium Phosphate Monobasic (Crystalline/Certified ACS), Fisher Chemical™
$333.67 - $19991.67
Chemical Identifiers
CAS | 7778-77-0 |
---|---|
Molecular Formula | H2KO4P |
Molecular Weight (g/mol) | 136.08 |
MDL Number | MFCD00011401 MFCD00147253 |
InChI Key | GNSKLFRGEWLPPA-UHFFFAOYSA-M |
Synonym | potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate |
PubChem CID | 516951 |
ChEBI | CHEBI:63036 |
IUPAC Name | potassium dihydrogen phosphate |
SMILES | [K+].OP(O)([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
P285-500
![]() |
Thermo Fisher Scientific
P285500 |
500 g | Poly Bottle |
Each for $333.67
Case of 6 Each for $2,002.02
|
![]() |
||||
P285-3
|
Thermo Fisher Scientific
P2853 |
3 kg | Poly Bottle |
Each for $1,101.92
Case of 4 Each for $4,407.68
|
|
||||
P285-10
|
Fisher Chemical
P28510 |
10 kg | Poly Pail |
Each for $3,077.92
|
|
||||
P285-50
|
Fisher Chemical
P28550 |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
7778-77-0 | |
136.08 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
516951 | |
potassium dihydrogen phosphate |
H2KO4P | |
MFCD00011401 MFCD00147253 | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
CHEBI:63036 | |
[K+].OP(O)([O-])=O |
Specifications
253°C | |
White | |
Solid | |
≥99 % | |
MFCD00011401 MFCD00147253 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
potassium dihydrogen phosphate | |
516951 | |
136.09 grams/ mol | |
Certified ACS | |
0.2% max. | |
>250°C |
7778-77-0 | |
4.5 | |
500 g | |
H2KO4P | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
[K+].OP(O)([O-])=O | |
136.08 | |
CHEBI:63036 | |
≥99% | |
Pass Test | |
Poly Bottle | |
Potassium Phosphate Monobasic |
Safety and Handling

CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:1
Flammability:0
Instability:1