missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium phosphate, tribasic, 97%, pure, anhydrous, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 387680010

 View more versions of this product

Catalog No. AC387680010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium phosphate, tribasic
Tribasic 97%, Anhydrous
White
1 kg
K3O4P
MFCD00036295
potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321
LWIHDJKSTIGBAC-UHFFFAOYSA-K
tripotassium;phosphate
62657
97%
Plastic bottle
1340.0°C
7778-53-2
Powder, Granules or Chunks
97%
K3PO4
15,7781
Solubility in water: 508g/L (25°C). Other solubilities: insoluble in alcohol
[O-]P(=O)([O-])[O-].[K+].[K+].[K+]
212.27
212.27
Pure

Chemical Identifiers

7778-53-2
212.27
LWIHDJKSTIGBAC-UHFFFAOYSA-K
62657
[O-]P(=O)([O-])[O-].[K+].[K+].[K+]
K3O4P
MFCD00036295
potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321
tripotassium;phosphate
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

GHS Signal Word: Danger

EINECSNumber : 231-907-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only