Learn More
Potassium phosphate, tribasic, 97%, pure, anhydrous
CAS: 7778-53-2 | K3O4P | 212.27 g/mol
Supplier: Thermo Scientific Chemicals 387680025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Potassium phosphate, tribasic | |
| Tribasic 97%, Anhydrous | |
| White | |
| 2.5 kg | |
| K3O4P | |
| MFCD00036295 | |
| potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321 | |
| LWIHDJKSTIGBAC-UHFFFAOYSA-K | |
| tripotassium;phosphate | |
| 62657 | |
| 97% | |
| Plastic bottle |
| 1340.0°C | |
| 7778-53-2 | |
| Powder, Granules or Chunks | |
| 97% | |
| K3PO4 | |
| 15,7781 | |
| Solubility in water: 508g/L (25°C). Other solubilities: insoluble in alcohol | |
| [O-]P(=O)([O-])[O-].[K+].[K+].[K+] | |
| 212.27 | |
| 212.27 | |
| Pure |
Chemical Identifiers
| 7778-53-2 | |
| 212.27 | |
| LWIHDJKSTIGBAC-UHFFFAOYSA-K | |
| 62657 | |
| [O-]P(=O)([O-])[O-].[K+].[K+].[K+] |
| K3O4P | |
| MFCD00036295 | |
| potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321 | |
| tripotassium;phosphate |
Safety and Handling
GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
EINECSNumber : 231-907-1
RUO – Research Use Only